2-(2-Thienyl)isonicotinsäure

Lieferant: COMBI-BLOCKS

YA-5955-1G
COBBYA-5955-1GEA 571 EUR
COBBYA-5955-1G
2-(2-Thienyl)isonicotinsäure
2-(2-Thienyl)isonicotinsäure
Formel: C₁₀H₇NO₂S
MDL: MFCD06496472
CAS-Nummer: 893723-57-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR