2-(3-Thienyl)isonicotinsäure

Lieferant: COMBI-BLOCKS

YA-5972-1G
COBBYA-5972-1GEA 0 EUR
COBBYA-5972-1G
2-(3-Thienyl)isonicotinsäure
2-(3-Thienyl)isonicotinsäure
Formel: C₁₀H₇NO₂S
MDL: MFCD06496474
CAS-Nummer: 893723-63-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR