2-(4-Fluorphenyl)isonicotinsäure

Lieferant: COMBI-BLOCKS

YA-6142-1G
COBBYA-6142-1GEA 0 EUR
COBBYA-6142-1G
2-(4-Fluorphenyl)isonicotinsäure
2-(4-Fluorphenyl)isonicotinsäure
Formel: C₁₂H₈FNO₂
MDL: MFCD14666481
CAS-Nummer: 883528-25-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR