2-Methyl-3-phenylbenzoesäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methyl-[1,1'-biphenyl]-3-carboxylic acid

YA-8643-1G
COBBYA-8643-1GEA 303 EUR
COBBYA-8643-1G
2-Methyl-3-phenylbenzoesäure
2-Methyl-3-phenylbenzoic acid
Formel: C₁₄H₁₂O₂
MDL: MFCD11554177
CAS-Nummer: 115363-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR