P3 SureLight® molekularbiologische Färbung
Lieferant: Columbia Biosciences
D7-000
COBSD7-000EA
560
EUR
COBSD7-000
P3 SureLight® molekularbiologische Färbung
P3 SureLight®
SureLight™ P3 is a proprietary dye can be used in prompt fluorescent detection, microplate applications, flow cytometry and protein microarrays.
- Application_NA Flow Cytometry, Cell-Based Assays, Microarrays and Microplate Applications
- Absorption max 614 nm
- Emission max 662 nm
Because its absorption and emission profile is similar to Cy5, it can be used in many of the same applications, however, it is 200 times brighter.
SureLight™ P3 has a linear structure that, on its largest diagonal, is about 80 nm long. It is comprised of C-PC and APC.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...