Needles, STERITEX®
Lieferant: CODAN
646008
CODA646008EA 106 EUR
CODA646008
Needles, STERITEX®
Injektionsnadeln
This needle is only used for the inlet of air into bottles.
  • Metal needle with non return valve
  • With 0,2 µm air filter
Bitte beachten Do not use for reconstitution of lyophilisate.