Infrared thermometer with laser sighting
Lieferant: COMARK
KM814
COMAKM814EA 324 EUR
COMAKM814
Infrared thermometer with laser sighting
Thermometer Digitalthermometer
The infrared HVAC thermometer with laser sighting is a pocket-sized infrared thermometer for fast and safe measurements with temperature range and accuracy ideally suited to general purpose HVAC applications.
  • Laser sighting to pinpoint target area
  • Back-lit LCD display, °C/°F button
  • Emissivity pre-set at 0,95, suitable for most organic materials and painted or oxidised surfaces
  • Hold function freezes displayed reading