Digital thermometer, N9005
Lieferant: COMARK
N9005
COMAN9005EA 983 EUR
COMAN9005
Digital thermometer, N9005
Thermometer Digitalthermometer
This industrial thermometer provide added flexibility and a sub-miniature probe connection.
  • High accuracy with rapid response
  • Permanent clock display
  • Countdown timer and alarm
  • Selectable automatic switch-Off
  • BioCote® antimicrobial protection
Can be used with thermocouple type K and T probes.
Lieferumfang: Supplied without probe.