Immersion probe
Lieferant: COMARK
PX33L
COMAPX33LEA 153 EUR
COMAPX33L
Immersion probe
Temperaturfühler
The PX33L Integral Plug Probe is a general purpose penetration probe that connects directly to a thermometer for single-handed temperature taking.
  • Temperature range: −40 to 150 °C / −40 to 302 °F
  • Sensor type: Thermistor (PST)
  • Thin tip: 3,3 mm
The PX33L Integral Plug Probe is fitted with a 6-Pin Lumberg connector with locking screw, to provide a strong and secure probe-to-instrument connection with better prevention of liquid ingress.
Not suitable for intrinsically safe applications