Digital clock/timer
Lieferant: COMARK
UTL882
COMAUTL882EA 140 EUR
COMAUTL882
Digital clock/timer
Timer Timer, digital
This kitchen clock has a countdown capacity of 24 hours. All of its three functions − clock, timer and countdown − can operate simultaneously.
  • Built-in memory allows storage of a pre-set time
  • Count up in 1 second resolution up to 24 hours
Standards: Compliant with HACCP (hazard analysis and critical control points) procedures.
Lieferumfang: Incl. magnet, pocket clip, stand and battery.