Conductivity standards, potassium chloride solutions
Lieferant: CONSORT
B560 B561 B562
CONSB560EA 23 EUR
CONSB560 CONSB561 CONSB562
Conductivity standards, potassium chloride solutions
Standards Leitfähigkeits-Standards
Potassium chloride solutions referenced at 25 °C.
  • Certified to 0,5%