Bucket system, 20 L
Lieferant: CONTEC
2769-KIT
CONT2769-KITEA 1390 EUR
CONT2769-KIT
Bucket system, 20 L
Mopps
Polypropylene bucket system is a stainless steel rolling cart with two polypropylene 20 L buckets, stainless steel drip cap, and angled stainless steel wringer.
  • The red and grey double bucket system allows for differentiation between cleaning and rinsing solutions
  • Compatible with all Contec® flat mops, including our PREMIRA™ advanced environmental hygiene system and Klean Max™
Durable, yet lightweight construction with four standard casters makes it easy to manoeuvre around pharmacy cleanrooms.