Softwall strip cleaner, DualClean™
Lieferant: CONTEC
2746S 2778 DCQT0001 DCMD0001 DCQT0002
CONT2746SEA 193 EUR
CONT2746S CONT2778 CONTDCQT0001 CONTDCMD0001 CONTDCQT0002
Softwall strip cleaner, DualClean™
Mopps
The DualClean™ Softwall Strip Cleaner is ideal for cleaning strip curtains and barriers in all environments from clean workspace enclosure to ISO Class 5 isolated cleanroom work cells.
This is a unique system that enables one operator to easily and quickly clean both sides for 4 to 12" plastic strip curtains in a single pass. The system consists of a fabric cover that is snugly installed over a stainless steel frame with two curved-surface arms. Each curtain strip to be cleaned is positioned between the two arms of the frame, with the arms contacting both sides of the curtain at once.More Product Information