PROSAT® Polynit Heatseal Wipes, 10% IPA/90% DIW
Lieferant: CONTEC
PSPS0048
CONTPSPS0048EA 1450 EUR
CONTPSPS0048
PROSAT® Polynit Heatseal Wipes, 10% IPA/90% DIW
Wischtücher Reinraum-Wischtücher Getränkte Reinraumwischtücher
These PROSAT® polynit heatseal wipes are presaturated with 10% semi-grade IPA/90% DI water.
  • Reduce particle and fiber generation
  • Abrasion- and chemical-resistant
  • Reduces VOC emissions and solvent use
  • Eliminates the need for in-house filtration
  • Meets USP grade purified water
PROSAT® Polynit heatseal wipes are manufactured from 100% knitted textured polyester with laser-cut sealed edges for very low levels of particles and fibers. Polyester wipes are chemically resistant and exceptionally low in particles and fibers. An interlock knit creates a durable fabric. The addition of a periodic additional stitch of a no-run interlock knit prevents the fabric from unraveling, creating an even more durable fabric with strong edges.