Contec® steriles denaturiertes Ethanol
Lieferant: CONTEC
SBC570DE SBT170DEW SBT0570DEW
CONTSBC570DEEA 321 EUR
CONTSBC570DE CONTSBT170DEW CONTSBT0570DEW
Contec® steriles denaturiertes Ethanol
Reinigungsmittel Desinfektionsmittel
Contec Sterile Denatured Ethanol is a blend of 70% v/v denatured ethanol with 30% water for injection or purified water. Contec’s ethanol is only denatured with IPA creating a very low residue product.
  • Guaranteed endotoxin levels of <0,25 EU/ml make it ideal for use in product contact areas
  • GMP manufactured under Grade A laminar flow air flow in a Grade C cleanroom ensures the alcohol, container and packaging are free from contamination and particulates
  • Trigger spray and 'bag in bottle' protected system pevents bottle contents from becoming contaminated
More Product Information