Cleaning swabs, CONSTIX® SC-9
Lieferant: CONTEC
SC-9
CONTSC-9EA 163 EUR
CONTSC-9
Cleaning swabs, CONSTIX® SC-9
Tupfer Reinigungstupfer
SC-9 round cotton swabs consist of high quality, long staple length cotton, tightly wrapped and shaped on one end of a high strength wooden handle.
  • Medium sized cotton ends, single-ended
  • High quality cotton fibres provide excellent particle entrapment and retention
  • Tightly wrapped head help maintain low levels of extractable residue
  • Heads are held to the handles using a methyl-cellulose binder
The high quality cotton fibres provide exceptional sorbency of most solvents and solutions. The heads are very tightly wrapped and shaped using a methyl-cellulose binder which minimises fibre shedding. SC series cotton swabs are absorbent to most common solvents including isopropyl alcohol and acetone and are ideal for use with aqueous-based solutions.
These swabs are Sanyo Huby brand swabs imported by Contec.
Verpackung: Swabs are packed 2000 per case.
Bitte beachten *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904