Cleaning swabs, sealed foam, CONSTIX® SF-5
Lieferant: CONTEC
SF-5
CONTSF-5EA 171 EUR
CONTSF-5
Cleaning swabs, sealed foam, CONSTIX® SF-5
Tupfer Reinigungstupfer
This sealed foam swab consists of a polyurethane foam bonded to a flex-tip PP handle.
  • Flexible swab head tip provides good surface contact and controlled pressure to surfaces
  • Polyurethane foam head provides good particle entrapment and removal
  • No adhesives or binders used
The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily sorbed for applications involving solvent delivery or removal. The SF-5 swab is excellent for applying resins and adhesives.
Though compatible with many common solvents, the SF-5 is not recommended for use with ketones such as acetone or MEK.
Verpackung: Swabs are packed 500 per case.
Bitte beachten *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904