Cleaning swabs, sealed foam, CONSTIX® SF-6
Lieferant: CONTEC
SF-6
CONTSF-6EA 356 EUR
CONTSF-6
Cleaning swabs, sealed foam, CONSTIX® SF-6
Tupfer Reinigungstupfer
The SF-6 sealed foam swab consists of a 100 PPI polyurethane foam adhesive bonded to a flexible nylon tip. The tip is mechanically secured within a tubular PP handle.
  • Flexible nylon tip allows for a softer application
  • Polyurethane foam head provides good particle entrapment for removal and solvent compatibility
The absorbant foam enables solvents, such as isopropanol and ethanol, to be readily sorbed for applications involving solvent delivery or removal.
Though compatible with many common solvents, the SF-6 is not recommended for use with ketones such as acetone or MEK.
Verpackung: Swabs are packed 500 per case.
Bitte beachten *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904