Cleaning swabs, sealed, CONSTIX® SP-11
Lieferant: CONTEC
SP-11
CONTSP-11EA 139 EUR
CONTSP-11
Cleaning swabs, sealed, CONSTIX® SP-11
Tupfer Reinigungstupfer
The SP-11 sealed polyester swab consists of 100% knitted polyester fabric that’s thermally bonded to a rigid PP handle.
  • Durable and absorbent
  • Features extremely low particles and residue levels
  • Polyester head provides enhanced sorbency, particle entrapment and removal
  • Polyester head makes the swab compatible with most common solvents, such as alcohols and ketones
  • Flexible tip enables access to very small areas and components
  • PP handle has a thin tip
The swab head is made of cleanroom laundered; double-layer knitted microfibre polyester material. The polyester head is thermally sealed and attached, also constructed without adhesives or binders.
Verpackung: Swabs are packed 500 per case.
Bitte beachten *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904