Cleanroom wipes, pre-saturated, sterile, SATWipes HydroPure
Lieferant: CONTEC
SSWC16HP
CONTSSWC16HPEA 1070 EUR
CONTSSWC16HP
Cleanroom wipes, pre-saturated, sterile, SATWipes HydroPure
Wischtücher Reinraum-Wischtücher Getränkte Reinraumwischtücher
Contec HydroPure is a blend of 6% hydrogen peroxide and purified water. Contec HydroPure is BPR authorised at EU level and efficacious against bacteria, fungi, moulds, and yeasts.
  • Impregnated wipes at point-of-use provide convenient and controlled application of disinfectant
  • Hydrogen Peroxide breaks down to water & oxygen leaving no residue
  • 100 polyester / cellulose wipes in a canister
  • Filtered to 0.2 microns in a Grade B cleanroom
  • Ideal for a Grade A / B areas
  • Not classed as corrosive
  • Double bagged in linear tear packaging
Contec HydroPure leaves little-to-no residue making it ideal for use in critical areas. Contec HydroPure is provided as a point-of-use SATWIPES system. Each double bagged pack contains a canister of 100 x 54gsm Amplitude Theta polyester/ cellulose dry wipes and a 500ml capped bottle of Contec HydroPure.
Simply add 500 ml of HydroPure to the dry roll, 5 mins before use, and use the impregnated wipes within 7 days.
Contec HydroPure is 0.2 micron filtered, and filled in a Grade A environment. The sterile product is filtered into pre-irradiated components. The dry wipe canister is irradiated at no less than 25 kGy
Bitte beachten Biozidprodukte vorsichtig verwenden. Vor Gebrauch stets Etikett und Produktinformationen lesen
*Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904