Polypropylene Wipes, SATWipes®
Lieferant: CONTEC
SWPP0004
CONTSWPP0004EA 749 EUR
CONTSWPP0004
Polypropylene Wipes, SATWipes®
Wischtücher Reinraum-Wischtücher Getränkte Reinraumwischtücher
Presaturated with IPA/DI water.
  • Rolls of wipes conveniently dispensed through a canister
  • 0.2 micron (µm) filtered provides a safe, consistent, and mild cleaning solution
  • Eliminates the need for solvent bottles to reduce solvent usage and reduces solvent use by up to 35%
SATWIPES® Polypropylene Wipes consist of a roll of (150) 152×230mm saturated wipes dispensed through a center-pull canister with a pop-up dispenser. The wipes are made of soft and clean non-woven polypropylene fabric. The solution is filtered to 0.2µm and provides a safe, consistent, and mild cleaning solution.
Eliminates the need for solvent bottles, reduces solvent handling and reduces solvent use by up to 35%. Refill rolls are available so the dispenser canister can be reused.