NUCLEIC-CARD™ Nucleic Acid Stabilisation and Archiving Devices
Lieferant: Copan
3001C50 3001C100 3005C100 3006C100 40U031D 40U032D
COPA3001C50EA 7220 EUR
COPA3001C50 COPA3001C100 COPA3005C100 COPA3006C100 COPA40U031D COPA40U032D
NUCLEIC-CARD™ Nucleic Acid Stabilisation and Archiving Devices
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
NUCLEIC-CARD™ is specifically designed to collect, transport, and store human DNA from buccal cells, saliva, blood, etc. Thanks to the unique chemical treatment, NUCLEIC-CARD™ preserves DNA for 20 years at room temperature.
  • Off-the-shelf products and custom formats: NUCLEIC-CARD™ is available in various off-the-shelf configurations (1,2,4 collection areas), clear or with a color indicator, as well as customisable sizes and layouts
  • Automation and cleaning area: NUCLEIC-CARD™ is compatible with CPA200™ and other punching systems
  • NUCLEIC-CARD™ includes an area for cleaning strikes while punching with manual or automated punchers
  • Traceability: every NUCLEIC-CARD™ is identified with a pre-printed barcode
  • Forensic DNA grade: ISO 18385:2016 compliance minimises the risk of detectable human nuclear DNA contamination in products used by the global forensic community
Direct amplification: the lysing treatment on the NUCLEIC-CARD™ allows a direct PCR STR analysis on a small punch of the card, without the need for the extraction step. NUCLEIC-CARD™ are not intended for diagnostic purposes. For collection, stabilization, processing, transport, and archiving of nucleic acid samples.
This product line groups a variety of product codes, each optimised to address specific needs.
Application: Long term sample storage
Standards: ISO 18385:2016. The validated EO treatment assesses the degradation of possible human DNA residuals. Free of amplifiable human DNA, detectable and amplifiable DNase, and RNase. Personnel STR database.
Bitte beachten The NUCLEIC-CARD™ is not intended for diagnostic purposes.