Polypropylene bottles, Corning®
Lieferant: Corning
1300P-1L 1300P-500W 1300P-500 1300P-250W 1300P-250 1300P-1LW
CORN1300P-1LEA 122.29 EUR
CORN1300P-1L CORN1300P-500W CORN1300P-500 CORN1300P-250W CORN1300P-250 CORN1300P-1LW
Polypropylene bottles, Corning®
Flaschen Spritzflaschen
Bottles are manufactured from low density polyethylene and are offered with a narrow or wide mouth design.
  • Hollow glass stopper style
  • Reusable
  • Non sterile
The cap and dispensing tube are manufactured from polypropylene to optimize backflow of the solution. The design permits minimal turbulence of the liquid in the bottle.