Media/storage bottles, wide mouth, round, with screw cap, PYREX®
Lieferant: Corning
1397-500
CORN1397-500EA 0 EUR
CORN1397-500
Media/storage bottles, wide mouth, round, with screw cap, PYREX®
Flaschen Zellkulturflaschen
Made of PYREX® borosilicate glass.
  • Resistant to thermal shock
  • Permanent white enamel graduations and marking spot
  • Traceability Code for easier documentation
  • Not recommended for vaccum use
Wide mouth media storage bottles are designed for heavy duty storage as well as mixing and sampling. They includes linerless, one-piece autoclavable orange polypropylene plug seal GLS80-threaded cap with a drip-free pouring ring.
Bitte beachten Do not tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage. These bottles should not be used in applications involving vacuum pressure as breakage may occur.