384-well microplate black/clear, round bottom, ultra-low attachment, spheroid, Corning®
Lieferant: Corning
3830BC
CORN3830BCEA 2540 EUR
CORN3830BC
384-well microplate black/clear, round bottom, ultra-low attachment, spheroid, Corning®
Mikrotiterplatten Zellkultur-Platten
Ultra-Low Attachment microplates feature a covalently bound hydrogel layer that effectively inhibits cellular attachment.
  • Sterilised by gamma irradiation
  • Opaque walls to prevent well-to-well crosstalk
  • Surface is noncytotoxic, biologically inert and nondegradable
  • With Generic Bar codes, and lid
Surface minimises protein absorption, enzyme activation and cellular activation; Black wall microplates have low background fluorescence, minimal light scatter and reduced crosstalk.
Can be used for both top and bottom reading instruments. Novel well geometryMore Product Information