96-well microplates, generic bar coded, Corning®
Lieferant: Corning
3897BC
CORN3897BCEA 439.82 EUR
CORN3897BC
96-well microplates, generic bar coded, Corning®
Mikrotiterplatten HTS-Mikrotiterplatten
A line of generic bar coded 96-well plates to better meet the demands of high throughput screening.
  • Labels applied to all 4 sides of the microplate to ensure usability regardless of scanner location
  • Each microplate is specially treated to reduce the impact of static build-up
  • Code 128 bar code format ensures compatibility with most bar code scanning and software systems
  • No sequence number duplicates
  • Repeatable ANSI grading of B or better
  • Exceptional print quality and resolution