Starter kit, Lambda™ Plus, Corning
Lieferant: Corning
4069
CORN4069EA 886.58 EUR
CORN4069
Starter kit, Lambda™ Plus, Corning
Pipetten Mikropipetten
The Lambda plus starter kit is designed for laboratories which are about to replace their old set of pipettors, and for customers who want to get a new set of pipettors which cover the most commonly used volumes.
The Lambda plus starter kit contains four Lambda Plus Single-Channel pipettors conveniently packaged with useful accessories (manual, calibration key, colour-coded rings, tube of piston grease and identification stickers).
Bestellinformation: This starter kit includes four Lambda Plus Single Channel pipettors in these volume sizes 0,5 to 10 µl, 2,0 to 20 µl, 20 to 200 µl, and 100 to 1000 µl and three racks of Corning IsoTip™ filtered pipette tips for the above sizes.