Counting chamber for corning cell counter, 0,1 mm, Corning®
Lieferant: Corning
480200
CORN480200EA 134.57 EUR
CORN480200
Counting chamber for corning cell counter, 0,1 mm, Corning®
Objektträger Zählkammern
The Corning counting chamber is a reusable glass cell counting slide for the Corning cell counter.
This gridless slide has a 0,1 mm depth same as a Neubauer Hemocytometer and comes standard with two high quality cover slips. These slides have a notch for easy loading of cell suspension.