Benchtop Workstation, Corning® Lambda™ EliteMax
Lieferant: Corning
6070
CORN6070EA 25860 EUR
CORN6070
Benchtop Workstation, Corning® Lambda™ EliteMax
Automatisierte Liquid-Handling-Systeme
The New Corning® LAMBDA™ EliteMax Semi-Automated Benchtop Pipettor is the optimal solution for purification, isolation, and extraction for sample prep and compound management workflows.
  • Blowout feature helps to dispense all excess liquid that remains in the tip after dispensing
  • Speed can be changed depending on the type of liquid or compound being transferred
  • The large touch screen controller allows the user to easily control the device
  • There is also an option for the instrument to be controlled manually on the touch screen controller
  • You can create multiple user accounts for storing preferences, protocols, and methods
The Corning® LAMBDA™ EliteMax Semi-Automated Benchtop Pipettor is a high throughout automation platform with a small footprint for:
- Plate filling from reagents reservoirs
- Serial dilutions, plate-to-plate transfers, tube-to-tube transfers, and plate replication
- Well-to-well transfers (cherry picking): Single-channel, 12-channel, 24-channel, and custom-channel heads are available
- Accurately and precisely transferring liquids and compounds between microplates
- Transferring by row or by column
Standards: Calibration certificate.
Verpackung: Palletized wooden crate.