Platform rockers, Corning® LSE™
Lieferant: Corning
6704 6706
CORN6704EA 936.21 EUR
CORN6704 CORN6706
Platform rockers, Corning® LSE™
Schüttler Rotations-/Wippschüttler
The Corning LSE Platform Rocker is a compact yet high capacity rocker that is ideal for western blotting and general mixing applications.
  • Rocker motion
  • Low profile and compact size
  • 11-pound load capacity
  • Non-slip rubber pads to prevent sample containers from moving during operation
  • Double platform model available for increased capacity
The adjustable rocking speed and fixed 7-degree tilt makes it optimal for staining fragile gels, as well as the more intensive action necessary for washing blots. The speed of the Corning Platform Rocker can beMore Product Information