Shaking incubators, 49 L, Corning® LSE™
Lieferant: Corning
6752
CORN6752EA 5241.96 EUR
CORN6752
Shaking incubators, 49 L, Corning® LSE™
Schüttler Inkubationsschüttler
This cabinet-style incubator has a compact footprint and can also be stacked to maximise space efficiency.
  • Interior made of mirrored stainless steel
  • Exterior made of cold rolled steel
The built in orbital shaker accepts a large assortment of platforms and clamps that can accommodate flasks, Petri dishes, and test tube racks. The maximum capacity is 4×1 L flasks.
Flask capacity: 5×500 ml, 9×250 ml, 16×125 ml.