Shaking incubators, Corning® LSE™
Lieferant: Corning
6754
CORN6754EA 5970.03 EUR
CORN6754
Shaking incubators, Corning® LSE™
Schüttler Inkubationsschüttler
This cabinet-style LSE™ shaking incubator is constructed with mirrored stainless steel interior and cold rolled steel exterior. Built in orbital shaker accepts a large assortment of platforms and clamps that can accommodate flasks, petri dishes and test tube racks. Mechanical convection provides a safe, stable temperature environment and fast chamber temperature recovery after opening the door.
  • Outstanding temperature uniformity
  • Glass door and internal light that allows easy viewing of samples without disturbing the chamber contents
  • Multiple shelving options
Maximum capacity is 4×2 L/6×1 L flasks.