Freezing Box
Lieferant: E3 Cortex
CARTISO15L
CORTCARTISO15LEA 192 EUR
CORTCARTISO15L
Freezing Box
Boxen Kühl- und Gefrierboxen, Kühl- und Gefrierbeutel
Isothermal box made of rigid polyurethane (PU) foam is allowing the shipment in low temperature (refrigeration or freezing) of products sensitive to thermal variations.
  • Requires the supply of a suitable source of cold, in sufficient quantity
It is particularly suitable for the transfer of products to maintain positive low temperature between 1 to 8 °C or negative less than -20 °C between 48 to 120 h.
Cooling source (solid carbon dioxide or bag or plate of eutectic gel) available in variable quantity according to the constraints relating to the product transported and the necessary transport time.