Magnetic stirring bars, glass-covered
Lieferant: COWIE
001.1219 001.1250
COWI001.1219EA 96.4 EUR
COWI001.1219 COWI001.1250
Magnetic stirring bars, glass-covered
Rührer Magnetrührstäbchen
Magnetic stirring bars designed for use with very abrasive media which may erode PTFE.
  • Cylindrical in shape with Alnico V magnetic core