Stirring shafts, plain
Lieferant: COWIE
005.100850EX 005.101050EX 005.101065EX 005.101250EX 005.101265EX 005.101275EX 005.1016100EX 005.1016750EX 005.1019750EX 005.1019100EX 005.100830EX 005.101030EX
COWI005.100850EXEA 70.7 EUR
COWI005.100850EX COWI005.101050EX COWI005.101065EX COWI005.101250EX COWI005.101265EX COWI005.101275EX COWI005.1016100EX COWI005.1016750EX COWI005.1019750EX COWI005.1019100EX COWI005.100830EX COWI005.101030EX
Stirring shafts, plain
Rührer Rührschäfte
Plain stirring shafts made of encapsulated stainless steel core with exposed end.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable