Dishes, evaporating, flat form
Lieferant: COWIE
006.1025 006.1050 006.1100 006.1180 006.1350
COWI006.1025EA 24.9 EUR
COWI006.1025 COWI006.1050 COWI006.1100 COWI006.1180 COWI006.1350
Dishes, evaporating, flat form
Schalen/Schüsseln/Töpfe Abdampfschalen
Made of PTFE.
  • Very good chemical resistance
  • Temperature resistant from -200 to +280 °C
  • Exceptionally smooth inner walls to prevent contamination
Thick walled, extremely robust.