Tubing connectors, T-type
Lieferant: COWIE
016.1104.9.2 016.1105.2 016.1102.2 016.1102.5.2 016.1103.2 016.1103.7.2 016.1104.2
COWI016.1104.9.2EA 27.1 EUR
COWI016.1104.9.2 COWI016.1105.2 COWI016.1102.2 COWI016.1102.5.2 COWI016.1103.2 COWI016.1103.7.2 COWI016.1104.2
Tubing connectors, T-type
Fittings und Verbinder
T-type connectors, made of PTFE, bayonet type, for tubings.
  • Very good chemical resistance
  • Very good temperature resistance