Standard valves
Lieferant: COWIE
016.1203.2 016.1204.2 016.1302.6.2 016.1303.2 016.1304.2 016.804.2 016.803.2 016.802.5.2 016.802.2 016.704.2 016.703.2 016.702.5.2 016.702.2 016.703.7.2 016.704.9.2 016.803.7.2 016.804.9.2 016.705.2 016.805.2 016.1202.6.2
COWI016.1203.2EA 51.1 EUR
COWI016.1203.2 COWI016.1204.2 COWI016.1302.6.2 COWI016.1303.2 COWI016.1304.2 COWI016.804.2 COWI016.803.2 COWI016.802.5.2 COWI016.802.2 COWI016.704.2 COWI016.703.2 COWI016.702.5.2 COWI016.702.2 COWI016.703.7.2 COWI016.704.9.2 COWI016.803.7.2 COWI016.804.9.2 COWI016.705.2 COWI016.805.2 COWI016.1202.6.2
Standard valves
Hähne
Standard valves made of PTFE.
  • Maximum Pressure 1 bar at 100 °C and 5 mm Hg
  • Valves may be sterlised at 135 °C
  • Very good chemical resistance