Spatulas
Lieferant: COWIE
043.120 043.150 043.180 043.210 043.240
COWI043.120EA 17.2 EUR
COWI043.120 COWI043.150 COWI043.180 COWI043.210 COWI043.240
Spatulas
Spatel
Double-ended spatula/scraper combination made of PTFE, with fine chamfered ends.