Self-retracting lifelines Ultra-Lok™, DBI-SALA®, 3M™
Lieferant: CAPITAL SAFETY
3504555
CPSD3504555EA 0 EUR
CPSD3504555
Self-retracting lifelines Ultra-Lok™, DBI-SALA®, 3M™
Fallschutz Verbindungsmittel und Seile
Ultra-Lok™ web and cable SRLs are rugged, highly engineered devices that can be counted on for user safety, efficiency and reliability. Available with synthetic rope, stainless steel or galvanised steel cable and aluminium or thermoplastic housing.
  • Anchor swivel
  • Lightweight and durable thermoplastic housing
  • Stainless steel components
  • Reserve lifeline
  • Semi-sealed design
  • Constant moving pawls, anti-ratchet system
  • Impact indicating hook
The RSQ™ model is the world's first dual-mode self-retracting lifeline for fall arrest an integrated rescue.
Capacity: 141 kg
Standards: EN 360