Portable fall arrest post, DBI-SALA, 3M™
Lieferant: CAPITAL SAFETY
8566691
CPSD8566691EA 0 EUR
CPSD8566691
Portable fall arrest post, DBI-SALA, 3M™
Fallschutz Höhensicherungsgeräte und Fallbegrenzer
The portable fall arrest post is a durable aluminium construction and specifically designed for use on top of transformers or other types of vertical platforms with potential fall hazards.
  • 3 independent swivel anchor points
  • Telescopic design of 832 to 1168 to 1454 mm
  • Levelling screws plumb system inclines up to 15°
  • Built-in anchor ring for horizontal lifeline attachment
  • Portable and fixed bases provide complete jobsite versatility
It is compact and lightweight design of only 17,20 kg makes the post very easy to transport and put in place. They can rotate 360° offering freedom of mobility for up to 3 workers with self-retracting lifelines or fixed length lanyards.
Standards: EN 795, Class B