D-ring anchorage, Protecta®, 3M™
Lieferant: CAPITAL SAFETY
AM211
CPSDAM211EA 0 EUR
CPSDAM211
D-ring anchorage, Protecta®, 3M™
Fallschutz Höhensicherungsgeräte und Fallbegrenzer
Protecta® anchorage D-ring made from 316 stainless steel; 12 mm fixing.
  • Can be used with three load directions
Standards: EN 795 Class A
Bestellinformation: Supplied without fixing.