Supelco® Organic Reference Standard, F.A.M.E. Mix C14 - C22
Lieferant: Merck
CRM18917
CRM18917EA
70.4
EUR
CRM18917
Supelco® Organic Reference Standard, F.A.M.E. Mix C14 - C22
Standards
Organische Standards
This certified reference material is useful for determining relative retention times and approximating response factors.
Bitte beachten P273 - P501