Food standards, Patulin solution, (CRM), TraceCERT®, Supelco®
Lieferant: Merck
CRM46914
CRM46914EA 143 EUR
CRM46914
Food standards, Patulin solution, (CRM), TraceCERT®, Supelco®
Standards Lebensmittel-Standards
Patulin is a mycotoxin produced by several mold species. It has been found in fruit, especially apples, fruit juice, cider and stewed fruits. Because of its highly toxic properties, this compound has been the focus of many studies.
Bitte beachten Storage temperature: Freezer −20 °C
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d488fe182c481fd27e303c241770595ccbd12148&vid=d488fe182c481fd27e303c241770595ccbd12148&prid=web&clid=SAPDAM