Aluminiumsulfat
Lieferant: CAESAR LORENTZ
|
Gefahr
|
Synonyme:
Aluminum sulphate
2072.0250
CSLO2072.0250EA
75.8
EUR
CSLO2072.0250
Aluminiumsulfat
Aluminium sulphate
Formel:
Al₂O₁₂S₃ Molecular Weight: 342,15 g/mol Schmelzpunkt: 770 °C Dichte: 2,672 g/cm³ (22.5 °C) |
MDL:
MFCD00003423 CAS-Nummer: 10043-01-3 EINECS: 233-135-0 Merck Index: 12,00381 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...