Magnesiumcitrat
Lieferant: CAESAR LORENTZ
Synonyme:
Trimagnesium dicitrate, Magnesium 2-hydroxypropane-1,2,3-tricarboxylate
2394-250G
CSLO2394-250GEA
37.5
EUR
CSLO2394-250G
Magnesiumcitrat
Magnesiumcitrat
Formel:
C₆H₈O₇·nMg |
MDL:
MFCD00036422 CAS-Nummer: 7779-25-1 EINECS: 231-923-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...