Lactose Monohydrat
Lieferant: CAESAR LORENTZ
Synonyme:
4-(β-D-Galactopyranosyl)-D-glucopyranose Monohydrat, 4-(β-D-Galactopyranosyl)-D-glucopyranose 1-Hydrat, 4-(β-D-Galactopyranosyl)-D-glucopyranose-1-Hydrat, 4-(β-D-Galactopyranosyl)-D-glucopyranose-Monohydrat
2582.0250
CSLO2582.0250EA
17.5
EUR
CSLO2582.0250
Lactose Monohydrat
Lactose Monohydrat
Formel:
C₁₂H₂₂O₁₁·1H₂O Molecular Weight: 360.32 g/mol Schmelzpunkt: 223 Lagertemperatur: Raumtemperatur |
CAS-Nummer:
10039-26-6 EINECS: 200-559-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...