Talk

Lieferant: CAESAR LORENTZ
Achtung

Synonyme: Magnesiumsilikat Monohydrat, Magnesiumsilikathydrat, Talkum

2660-1KG
CSLO2660-1KGEA 25.7 EUR
CSLO2660-1KG
Talk
Talk
Formel: Mg₃O₁₁Si₄·1H₂O
Molecular Weight: 379,27 g/mol
Schmelzpunkt: >1200 °C
Dichte: 2,58…2.83 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00792903
CAS-Nummer: 14807-96-6
EINECS: 238-877-9
Merck Index: 13,09127

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR