Sonnenblumenkernöl

Lieferant: CAESAR LORENTZ

7309-1L 7309-5L
CSLO7309-1LEA 44.2 EUR
CSLO7309-1L CSLO7309-5L
Sonnenblumenkernöl
Sunflower seed oil
MDL: MFCD00132403
CAS-Nummer: 8001-21-6
EINECS: 232-273-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR