Weizenkeimöl

Lieferant: CAESAR LORENTZ

7723.0100
CSLO7723.0100EA 24.6 EUR
CSLO7723.0100
Weizenkeimöl
Wheat germ oil
MDL: MFCD00132584
CAS-Nummer: 8006-95-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR